CAS RN 92478-27-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Morinidazole, MedChemExpress
MedChemExpress Morinidazole is a novel 5-nitroimidazole antimicrobial drug that undergoes extensive metabolism in humans via N+-glucuronidation and sulfation, for the treatment of bacterial infections including appendicitis and pelvic inflammatory disease (PID) caused by anaerobic bacteria.