CAS RN 92169-45-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Neuromedin N
MedChemExpress Neuromedin N is a potent modulator of dopamine D2 receptor agonist binding in rat neostriatal membranes.
2
MedChemExpress Neuromedin N
MedChemExpress Neuromedin N is a potent modulator of dopamine D2 receptor agonist binding in rat neostriatal membranes.
3
MedChemExpress Neuromedin N
MedChemExpress Neuromedin N is a potent modulator of dopamine D2 receptor agonist binding in rat neostriatal membranes.