CAS RN 920509-32-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Resmetirom, MedChemExpress
MedChemExpress Resmetirom (MGL-3196) is a highly selective thyroid hormone receptor β (THR-β) agonist with an EC50 value of 0.21 μM.