CAS RN 919486-40-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ AS 1517499
STAT6 inhibitor
2
AS1517499, MedChemExpress
MedChemExpress AS1517499 is a potent and brain-permeable STAT6 phosphorylation inhibitor with an IC50 of 21 nM.