CAS RN 918505-61-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BRAF inhibitor, MedChemExpress
MedChemExpress BRAF inhibitor is a B-Raf inhibitor extracted from patent WO/2011103196 A1, Compound P-0850.