CAS RN 91757-46-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSC 601980 (analog), MedChemExpress
MedChemExpress NSC 601980 analog is the analog of the NSC601980, NSC601980 shows antitumor activity in the yeast screening experiment, which can inhibit cell proliferation in the COLO 205 and HT29 with Log GI 50 of -6.6 and -6.9 respectively.