CAS RN 917497-70-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SC144 hydrochloride, MedChemExpress
MedChemExpress SC144 hydrochloride is a first-in-class, orally active gp130 (IL6-beta) inhibitor.