CAS RN 916918-84-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ansofaxine hydrochloride, MedChemExpress
MedChemExpress Ansofaxine hydrochloride (LY03005; LPM570065) is a triple reuptake inhibitor; inhibits serotonin, dopamine and norepinephrine reuptake with IC50 values of 723, 491 and 763 nM, respectively.