CAS RN 915191-42-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MF498, MedChemExpress
MedChemExpress MF498 is a novel and selective E prostanoid receptor 4 (EP4 receptor) antagonist, displayed strong binding affinity for the EP4 receptor with Ki of 0.7 nM.