CAS RN 912287-56-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YHO-13177, MedChemExpress
MedChemExpress YHO-13177 is a potent and specific inhibitor of BCRP; potentiated the cytotoxicity of SN-38 in cancer cells and no effect on P-glycoprotein-mediated paclitaxel resistance in MDR1-transduced human leukemia K562 cells.