CAS RN 911714-45-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SP-420, MedChemExpress
MedChemExpress SP-420 is a desferrithiocin analogue with iron-clearing efficiency with ICE value of 26.7; more potent than desferrithiocin.