CAS RN 91037-75-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress GRGDSP
MedChemExpress GRGDSP, a synthetic linear RGD peptide, is an integrin inhibitor.
2
MedChemExpress GRGDSP
MedChemExpress GRGDSP, a synthetic linear RGD peptide, is an integrin inhibitor.