CAS RN 905579-51-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ MLN 4924
Potent and selective NEDD8 activating enzyme (NAE) inhibitor; cell permeable
3
Pevonedistat, MedChemExpress
MedChemExpress Pevonedistat (MLN4924) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor with an IC50 of 4.7 nM.
4