CAS RN 9050-30-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Heparan Sulfato (grado técnico), TRC
CAS: 9050-30-0 Fórmula molecular: C26H45N3O39S6 Peso molecular (g/mol): 1216 Sinónimo: Alpha-idosane,HHS 5,Heparan N-Sulfate,Heparan Sulfate,Heparan Sulphate,Heparitin,Heparitin Monosulfate,N-Acetylheparan Sulfate,Suleparoid,Tavidan;
2
Heparan Sulfate, MedChemExpress
MedChemExpress Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.