CAS RN 902156-99-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NVP-LCQ195, MedChemExpress
MedChemExpress NVP-LCQ195 (AT9311; LCQ195) is a small molecule heterocyclic inhibitor of CDK1, CDK2, CDK3 and CDK5 with IC50 of 1-42 nM.