CAS RN 901751-47-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
iCRT3, MedChemExpress
MedChemExpress iCRT3 is an inhibitor of both Wnt and β-catenin-responsive transcription.