CAS RN 9014-63-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Xylan, TRC
CAS: 9014-63-5 Fórmula molecular: C5H10O6 Peso molecular (g/mol): 166.13 Sinónimo: Xylans,CMT 85,D-Xylan,DP 42 Nombre IUPAC: (2R,3R,5S,6S)-tetrahidropiran-2,3,4,5,6-pentol SMILES: O[C@@H]1O[C@H](O)[C@@H](O)C(O)[C@H]1O
2
Xylan, MedChemExpress
MedChemExpress Xylan represents the main hemicellulose component in the secondary plant cell walls of flowering plants. Xylan is a polysaccharide made from units of xylose and contains predominantly β-D-xylose units linked as in cellulose.