CAS RN 9012-54-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Cellulase
MedChemExpress Cellulase is an enzyme catalyzing the hydrolysis of certain linkages in cellulose and other carbohydrates.
2
MedChemExpress Cellulase
MedChemExpress Cellulase is an enzyme catalyzing the hydrolysis of certain linkages in cellulose and other carbohydrates.