CAS RN 898800-26-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AZ876, MedChemExpress
MedChemExpress AZ876 is a potent and high-affinity LXR agonist. AZ876 displays 25-fold and 2.5-fold more potent than GW3965 (HY-10627) on human (h)LXRα and hLXRβ respectively.