CAS RN 898280-07-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
XL228, MedChemExpress
MedChemExpress XL228 is a multi-targeted tyrosine kinase inhibitor with IC50s of 5, 3.1, 1.6, 6.1, 2 nM for Bcr-Abl, Aurora A, IGF-1R, Src and Lyn, respectively.