CAS RN 898230-59-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Morinidazole (R enantiomer), MedChemExpress
MedChemExpress Morinidazole R enantiomer is the R-enantiomer of Morinidazole. Morinidazole is a new 5-nitroimidazole class antimicrobial agent. Morinidazole R enantiomer is the less active enantiomer.