CAS RN 896705-16-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ BMH 21
RNA polymerase I inhibitor; also p53 pathway activator
2
BMH-21, MedChemExpress
MedChemExpress BMH-21 is a first-in-class DNA intercalator which inhibits RNA polymerase I (Pol I) transcription. BMH-21 possesses anticancer activity.