CAS RN 894002-50-7
IUPAC Nombre:
3-{[(2-methylphenyl)methyl]sulfanyl}-6-(pyridin-2-yl)pyridazine
Sinónimos:
3-{[(2-methylphenyl)methyl]sulfanyl}-6-(pyridin-2-yl)pyridazine
Peso molecular (g/mol):
293.39
Fórmula molecular:
C17H15N3S
InChi Key:
DUUQLWDHNYFUPP-UHFFFAOYSA-N
SMILES:
CC1=CC=CC=C1CSC1=NN=C(C=C1)C1=CC=CC=N1
1
–
1
de
1
resultados
1
LDN-212320, MedChemExpress
MedChemExpress LDN-212320 (LDN-0212320) is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator (at translational level). LDN-212320 (LDN-0212320) prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC[1].