CAS RN 890190-22-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
WDR5-0103, MedChemExpress
MedChemExpress WDR5-0103 is a potent and selective WD repeat-containing protein 5 (WDR5) antagonist with Kd of 450 nM.