CAS RN 889665-86-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Cloruro de epiberberina, TRC
CAS: 889665-86-5 Fórmula molecular: C20 H18 N O4 . Cl Peso molecular (g/mol): 371.81 Sinónimo: 7,8,13,13a-Tetradehydro-2,3-dimethoxy-9,10-(methylenedioxy)berbinium Chloride SMILES: [Cl-].COc1cc2CC[n+]3cc4c5OCOc5ccc4cc3c2cc1OC
2
Epiberberine chloride, MedChemExpress
MedChemExpress Epiberberine chloride is an alkaloid isolated from Coptis chinensis, acts as a potent AChE and BChE inhibitor, and a non-competitive BACE1 inhibitor, with IC50s of 1.07, 6.03 and 8.55 μM, respectively.