CAS RN 887650-05-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Lyn-IN-1, MedChemExpress
MedChemExpress Lyn-IN-1 (Bafetinib analog) is a potent and selective dual Bcr-Abl/Lyn inhibitor, extracted from patent WO2014169128A1.