CAS RN 886992-99-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ac-Gly-BoroPro, MedChemExpress
MedChemExpress Ac-Gly-BoroPro is a selective FAP inhibitor with a Ki of 23 nM.