CAS RN 88678-31-3
IUPAC Nombre:
N-(6-methoxypyridin-2-yl)-N-methyl-1-(5,6,7,8-tetrahydronaphthalen-2-yloxy)methanethioamide
Sinónimos:
liranaftate 利拉萘酯 liranaftato piritetrate zefnart liranaftatum
Peso molecular (g/mol):
328.43
Fórmula molecular:
C18H20N2O2S
InChi Key:
VPHPQNGOVQYUMG-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=N1)N(C)C(=S)OC1=CC=C2CCCCC2=C1
1
–
1
de
1
resultados
1
Liranaftate, MedChemExpress
MedChemExpress Liranaftate (Piritetrate) is a squalene epoxidase inhibitor with anti-fungicidal activities. Liranaftate can be used for the research of dermatophytes. Liranaftate also suppresses fungal element-promoted production of IL-8 and experimental inflammation.