CAS RN 885220-61-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Verucerfont, MedChemExpress
MedChemExpress Verucerfont is a corticotropin-releasing factor receptor 1 (CRF1) antagonist with IC50s of ∼6.1, >1000 and >1000 nM for CRF1, CRF2, and CRF-BP, respectively.