CAS RN 882257-11-6
IUPAC Nombre:
1-{5-[(2,3-dichlorophenyl)sulfanyl]-4-nitrothiophen-2-yl}ethan-1-one
Sinónimos:
1-{5-[(2,3-dichlorophenyl)sulfanyl]-4-nitrothiophen-2-yl}ethanone
Peso molecular (g/mol):
348.21
Fórmula molecular:
C12H7Cl2NO3S2
InChi Key:
LKZLGMAAKNEGCH-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(=C(SC2=C(Cl)C(Cl)=CC=C2)S1)[N+]([O-])=O
1
–
1
de
1
resultados
1
P005091, MedChemExpress
MedChemExpress P005091 is a selective and potent inhibitor of ubiquitin-specific protease 7 (USP7) with an EC50 of 4.2 μM.