CAS RN 879085-55-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Novus Biologicals™ Vismodegib Hedgehog inhibitor
May be dissolved in DMSO (200 mg/mL); or ethanol (10 mg/mL, warm)
2
Tocris Bioscience™ Vismodegib
Potent hedgehog pathway inhibitor
3
Vismodegib, MedChemExpress
MedChemExpress Vismodegib (GDC-0449) is an orally active hedgehog pathway inhibitor with an IC50 of 3 nM. Vismodegib also inhibits P-gp, ABCG2 with IC50 values of 3.0 μM and 1.4 μM, respectively.