CAS RN 878451-87-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ RS 246204
R-Spondin-1 substitute used to initiate small intestinal organoids
2
RS-246204, MedChemExpress
MedChemExpress RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein.