CAS RN 876310-60-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
RF9, MedChemExpress
MedChemExpress RF9 is a potent and selective Neuropeptide FF receptor antagonist, with Ki values of 58 and 75 nM for hNPFF1R and hNPFF2R, respectively.