CAS RN 872201-12-2
IUPAC Nombre:
4-(2-{3-bromo-4-[2-(4-hydroxyphenyl)ethenyl]phenyl}ethenyl)phenol
Sinónimos:
4-(2-{3-bromo-4-[2-(4-hydroxyphenyl)ethenyl]phenyl}ethenyl)phenol
Peso molecular (g/mol):
393.28
Fórmula molecular:
C22H17BrO2
InChi Key:
OXPHQQMZTXMEGO-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=CC2=CC(Br)=C(C=CC3=CC=C(O)C=C3)C=C2)C=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ K 114
CAS: 872201-12-2 Fórmula molecular: C22H17BrO2
2
MedChemExpress K114
MedChemExpress K114, a fluorescent Congo Red analogue, binds tightly to amyloid fibrils with an EC50 of 20-30 nM. K114 is an efficient detector of semen-derived enhancer of virus infection (SEVI).