CAS RN 87096-84-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Neuromedin B
MedChemExpress Neuromedin B (NMB) is a member of Bombesin (BN)-like peptide family in mammals.
2
MedChemExpress Neuromedin B
MedChemExpress Neuromedin B (NMB) is a member of Bombesin (BN)-like peptide family in mammals.
3
MedChemExpress Neuromedin B
MedChemExpress Neuromedin B (NMB) is a member of Bombesin (BN)-like peptide family in mammals.
4
MedChemExpress Neuromedin B
MedChemExpress Neuromedin B (NMB) is a member of Bombesin (BN)-like peptide family in mammals.