CAS RN 870087-36-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Apilimod dimesylate
Potent and selective PIKfyve inhibitor
2
Apilimod mesylate, MedChemExpress
MedChemExpress Apilimod (STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC50s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. Apilimod is a potent and highly selective PIKfyve inhibitor.