CAS RN 869363-13-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MLN8054, MedChemExpress
MedChemExpress MLN8054 is a potent, selective and orally available aurora A kinase inhibitor with an IC50 of 4 nM.