CAS RN 86764-11-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Isoastragaloside II, MedChemExpress
MedChemExpress Isoastragaloside II is an astragaloside, which is isolated from the hairy root culture of Astragalus membranaceus.