CAS RN 863238-55-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Salicyl-AMS, MedChemExpress
MedChemExpress Salicyl-AMS is a mycobactin biosynthesis inhibitor which can also inhibit M. tuberculosis growth in vitro under iron-limited conditions.