CAS RN 858668-07-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EMD534085, MedChemExpress
MedChemExpress EMD534085 is a potent and selective inhibitor of the mitotic kinesin-5 with an IC50 of 8 nM.