CAS RN 858474-14-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSC23925, MedChemExpress
MedChemExpress NSC23925 is a novel, selective and effective P-glycoprotein (Pgp) inhibitor.