CAS RN 858102-78-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
E-982, MedChemExpress
MedChemExpress E-982 is a steroid used for the on-line screening of the DNA unwinding element binding protein (DUE-B) immobilized protein column.