CAS RN 857402-63-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Retaspimycin Hydrochloride, MedChemExpress
MedChemExpress Retaspimycin Hydrochloride is a potent inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9.