CAS RN 85371-64-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Novus Biologicals™ Pinacidil Monohydrate K+ channel activator
K+ channel activator
2
Pinacidil monohydrate, MedChemExpress
MedChemExpress Pinacidil (P-1134) monohydrate, an antihypertensive drug, is a potassium channel activator.