CAS RN 852391-15-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Necrostatin 2 racemate, MedChemExpress
MedChemExpress Necrostatin 2 racemate (Nec-1S), the Nec-1 stable, is a potent and specific RIPK1 inhibitor lacking the IDO-targeting effect.