CAS RN 851916-42-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MK-0812 Succinate, MedChemExpress
MedChemExpress MK-0812 Succinate is a potent and selective CCR2 antagonist with high affinity at CCR2.