CAS RN 850664-21-0
IUPAC Nombre:
N-(3-{[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-1H-imidazo[4,5-c]pyridin-6-yl]oxy}phenyl)-4-[2-(morpholin-4-yl)ethoxy]benzamide
Sinónimos:
N-(3-{[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-6-yl]oxy}phenyl)-4-[2-(morpholin-4-yl)ethoxy]benzamide
Peso molecular (g/mol):
570.61
Fórmula molecular:
C29H30N8O5
InChi Key:
YOVNFNXUCOWYSG-UHFFFAOYSA-N
SMILES:
CCN1C2=CC(OC3=CC=CC(NC(=O)C4=CC=C(OCCN5CCOCC5)C=C4)=C3)=NC=C2N=C1C1=NON=C1N
1
–
2
de
2
resultados
1
Tocris Bioscience™ GSK 269962
CAS: 850664-21-0 Fórmula molecular: C29H30N8O5
2
GSK269962A, MedChemExpress
MedChemExpress GSK269962A (GSK 269962) is a potent ROCK inhibitor with IC50s of 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively. GSK269962A has anti-inflammatory and vasodilatory activities.