CAS RN 848217-00-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
H4 Receptor antagonist 1, MedChemExpress
MedChemExpress H4 Receptor antagonist 1 is a potent and selective histamine H4 receptor inverse agonist, with an IC50 of 19 nM.