CAS RN 848193-68-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(S)-Selisistat, MedChemExpress
MedChemExpress (S)-Selisistat ((S)-EX-527) is a potent and selective SIRT1 inhibitor, with an IC50 of 98 nM.