CAS RN 844-26-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bithionol sulfoxide, MedChemExpress
MedChemExpress Bithionol sulfoxide(Bitin-S) is a clinically approved anti-parasitic drug; has been shown to inhibit solid tumor growth in several preclinical cancer models.